The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271851.1 | NP_001258780.1 | 2 - 3 | 510 | 108 |
NM_022157.3 | NP_071440.1 | 2 - 3 | 612 | 108 | |
GenBank mRNA | AF272035.1 | - | 2 - 3 | 440 | 108 |
AF323609.1 | - | 2 - 3 | 435 | 108 | |
AK298573.1 | - | 1 - 2 | 135 | 108 | |
AK315791.1 | - | 2 - 3 | 537 | 108 | |
BC016668.1 | - | 2 - 3 | 457 | 108 | |
BG717489.1 | - | 2 - 3 | 620 | 108 |