The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297655.1 | NP_001284584.1 | 7 - 8 | 754 | 83 |
NM_001297656.1 | NP_001284585.1 | 7 - 8 | 995 | 83 | |
NM_001297657.1 | NP_001284586.1 | 7 - 8 | 682 | 83 | |
NM_006845.3 | NP_006836.2 | 8 - 9 | 877 | 83 | |
XM_011540540.2 | XP_011538842.1 | 8 - 9 | 1124 | 83 | |
XM_011540541.2 | XP_011538843.1 | 7 - 8 | 1065 | 83 | |
GenBank mRNA | AB264115.1 | - | 8 - 9 | 877 | 83 |
AK290688.1 | - | 8 - 9 | 803 | 83 | |
AK300756.1 | - | 7 - 8 | 711 | 83 | |
AK302267.1 | - | 7 - 8 | 650 | 83 | |
AK312606.1 | - | 8 - 9 | 833 | 83 | |
AY026505.1 | - | 7 - 8 | 948 | 83 | |
BC008764.2 | - | 8 - 9 | 816 | 83 | |
BC014924.1 | - | 8 - 9 | 814 | 83 | |
BT006759.1 | - | 8 - 9 | 762 | 83 | |
CR450302.1 | - | 8 - 9 | 762 | 83 | |
U63743.1 | - | 8 - 9 | 816 | 83 |