Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPEP2
Entrez Gene ID
64174
Gene Name
dipeptidase 2
Gene Aliases
MBD2
Chromosome Location
Chr.16: 67987390 - 68000635 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022355.3 NP_071750.1 2 - 3 455 76
NR_136706.1 3 - 4 932 76
XM_005256090.4 XP_005256147.2 1 - 2 1412 76
XM_011523266.1 XP_011521568.1 2 - 3 398 76
XM_011523268.2 XP_011521570.1 1 - 2 1412 76
XM_011523271.2 XP_011521573.1 1 - 2 1412 76
XM_017023542.1 XP_016879031.1 1 - 2 1412 76
GenBank mRNA AJ295149.1 - 2 - 3 390 76
AK092884.1 - 3 - 4 932 76
AK315090.1 - 1 - 2 432 76
AY358454.1 - 1 - 2 216 76
BP299049.1 - 2 - 3 435 76
DA677374.1 - 2 - 3 398 76
DB038910.1 - 1 - 2 448 76

View Full Product Details