Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8A
Entrez Gene ID
731
Gene Name
complement component 8 alpha subunit
Gene Aliases
-
Chromosome Location
Chr.1: 56854770 - 56918221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000562.2 NP_000553.1 3 - 4 450 66
XM_011542079.2 XP_011540381.1 3 - 4 424 66
XM_017002234.1 XP_016857723.1 3 - 4 424 66
GenBank mRNA AK309596.1 - 3 - 4 424 66
BC020702.1 - 3 - 4 423 66
BC132911.1 - 3 - 4 342 66
BC132913.1 - 3 - 4 342 66
M16974.1 - 3 - 4 455 66

View Full Product Details