The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292032.1 | NP_001278961.1 | 1 - 2 | 123 | 80 |
NM_003427.4 | NP_003418.2 | 1 - 2 | 123 | 80 | |
XM_011514846.2 | XP_011513148.1 | 1 - 2 | 97 | 80 | |
XM_011514849.2 | XP_011513151.1 | 1 - 2 | 107 | 80 | |
XM_011514854.2 | XP_011513156.1 | 1 - 2 | 107 | 80 | |
XM_017011254.1 | XP_016866743.1 | 1 - 2 | 107 | 80 | |
GenBank mRNA | AK294605.1 | - | 1 - 2 | 107 | 80 |
BC000133.2 | - | 1 - 2 | 103 | 80 | |
BC002549.2 | - | 1 - 2 | 104 | 80 | |
DC299240.1 | - | 1 - 2 | 123 | 80 | |
M91592.1 | - | 1 - 2 | 62 | 80 |