The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284351.2 | NP_001271280.1 | 5 - 6 | 1050 | 94 |
NM_004174.3 | NP_004165.2 | 5 - 6 | 1050 | 94 | |
GenBank mRNA | BC011555.1 | - | 3 - 4 | 585 | 94 |
BC101669.1 | - | 5 - 6 | 946 | 94 | |
BC101671.1 | - | 5 - 6 | 946 | 94 | |
BC143328.1 | - | 5 - 6 | 946 | 94 | |
U28043.1 | - | 5 - 6 | 948 | 94 |