The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287758.1 | NP_001274687.1 | 30 - 31 | 3116 | 122 |
NM_001287759.1 | NP_001274688.1 | 29 - 30 | 3065 | 122 | |
NM_001287760.1 | NP_001274689.1 | 29 - 30 | 3065 | 122 | |
NM_001847.3 | NP_001838.2 | 29 - 30 | 2960 | 122 | |
NM_033641.3 | NP_378667.1 | 29 - 30 | 3065 | 122 | |
XM_006724617.3 | XP_006724680.1 | 30 - 31 | 3005 | 122 | |
XM_011530852.2 | XP_011529154.1 | 30 - 31 | 3005 | 122 | |
XM_011530853.2 | XP_011529155.1 | 28 - 29 | 3721 | 122 | |
XM_011530854.2 | XP_011529156.1 | 30 - 31 | 3006 | 122 | |
GenBank mRNA | BC140855.1 | - | 29 - 30 | 2961 | 122 |
BC144669.1 | - | 29 - 30 | 2961 | 122 | |
CA396616.1 | - | 4 - 5 | 576 | 122 | |
U04845.1 | - | 29 - 30 | 2943 | 122 |