The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287758.1 | NP_001274687.1 | 32 - 33 | 3422 | 61 |
NM_001287759.1 | NP_001274688.1 | 31 - 32 | 3371 | 61 | |
NM_001287760.1 | NP_001274689.1 | 31 - 32 | 3371 | 61 | |
NM_001847.3 | NP_001838.2 | 31 - 32 | 3266 | 61 | |
NM_033641.3 | NP_378667.1 | 31 - 32 | 3371 | 61 | |
XM_006724617.3 | XP_006724680.1 | 32 - 33 | 3311 | 61 | |
XM_011530852.2 | XP_011529154.1 | 32 - 33 | 3311 | 61 | |
XM_011530853.2 | XP_011529155.1 | 30 - 31 | 4027 | 61 | |
XM_011530854.2 | XP_011529156.1 | 32 - 33 | 3312 | 61 | |
GenBank mRNA | BC140855.1 | - | 31 - 32 | 3267 | 61 |
BC144669.1 | - | 31 - 32 | 3267 | 61 | |
CA489342.1 | - | 1 - 2 | 167 | 61 | |
D21337.1 | - | 31 - 32 | 3371 | 61 | |
U04845.1 | - | 31 - 32 | 3249 | 61 |