Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL4A6
Entrez Gene ID
1288
Gene Name
collagen type IV alpha 6 chain
Gene Aliases
CXDELq22.3, DELXq22.3, DFNX6
Chromosome Location
Chr.X: 108155607 - 108439497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287758.1 NP_001274687.1 7 - 8 746 73
NM_001287759.1 NP_001274688.1 7 - 8 746 73
NM_001287760.1 NP_001274689.1 7 - 8 746 73
NM_001847.3 NP_001838.2 7 - 8 641 73
NM_033641.3 NP_378667.1 7 - 8 746 73
XM_006724617.3 XP_006724680.1 7 - 8 635 73
XM_011530852.2 XP_011529154.1 7 - 8 635 73
XM_011530853.2 XP_011529155.1 5 - 6 1351 73
XM_011530854.2 XP_011529156.1 7 - 8 636 73
GenBank mRNA AW082083.1 - 7 - 8 73
BC000874.1 - 7 - 8 680 73
BC140855.1 - 7 - 8 642 73
BC144669.1 - 7 - 8 642 73
D21337.1 - 7 - 8 746 73

View Full Product Details