Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF76
Entrez Gene ID
7629
Gene Name
zinc finger protein 76
Gene Aliases
D6S229E, ZNF523, Zfp523
Chromosome Location
Chr.6: 35259473 - 35295987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292032.1 NP_001278961.1 3 - 4 378 73
NM_003427.4 NP_003418.2 3 - 4 378 73
XM_011514846.2 XP_011513148.1 3 - 4 352 73
XM_011514847.2 XP_011513149.1 3 - 4 490 73
XM_011514849.2 XP_011513151.1 3 - 4 362 73
XM_011514850.2 XP_011513152.1 3 - 4 490 73
XM_011514854.2 XP_011513156.1 3 - 4 362 73
XM_011514855.2 XP_011513157.1 3 - 4 490 73
XM_017011248.1 XP_016866737.1 3 - 4 490 73
XM_017011249.1 XP_016866738.1 3 - 4 490 73
XM_017011250.1 XP_016866739.1 3 - 4 490 73
XM_017011251.1 XP_016866740.1 3 - 4 490 73
XM_017011252.1 XP_016866741.1 3 - 4 490 73
XM_017011253.1 XP_016866742.1 3 - 4 490 73
XM_017011254.1 XP_016866743.1 3 - 4 362 73
XM_017011255.1 XP_016866744.1 3 - 4 490 73
XM_017011256.1 XP_016866745.1 3 - 4 505 73
XM_017011257.1 XP_016866746.1 3 - 4 490 73
GenBank mRNA AB208784.1 - 3 - 4 655 73
AK294605.1 - 3 - 4 362 73
AK302903.1 - 3 - 4 480 73
AK304626.1 - 3 - 4 729 73
BC000133.2 - 3 - 4 358 73
BC002549.2 - 3 - 4 359 73
BT007316.1 - 2 - 3 154 73
DC299240.1 - 3 - 4 378 73
M91592.1 - 3 - 4 317 73

View Full Product Details