The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292032.1 | NP_001278961.1 | 11 - 12 | 1555 | 65 |
XM_017011251.1 | XP_016866740.1 | 11 - 12 | 1703 | 65 | |
XM_017011252.1 | XP_016866741.1 | 11 - 12 | 1703 | 65 | |
XM_017011253.1 | XP_016866742.1 | 11 - 12 | 1667 | 65 | |
XM_017011254.1 | XP_016866743.1 | 11 - 12 | 1539 | 65 | |
XM_017011256.1 | XP_016866745.1 | 11 - 12 | 1682 | 65 | |
XM_017011257.1 | XP_016866746.1 | 11 - 12 | 1680 | 65 | |
GenBank mRNA | M91592.1 | - | 11 - 12 | 1494 | 65 |