The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017947.2 | NP_060417.2 | 11 - 12 | 2194 | 70 |
GenBank mRNA | AK000740.1 | - | 11 - 12 | 2192 | 70 |
AK222886.1 | - | 11 - 12 | 2181 | 70 | |
AL834481.1 | - | 9 - 10 | 1890 | 70 | |
BC012079.1 | - | 11 - 12 | 2184 | 70 |