The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017947.2 | NP_060417.2 | 1 - 2 | 165 | 55 |
GenBank mRNA | AK000740.1 | - | 1 - 2 | 163 | 55 |
AK222886.1 | - | 1 - 2 | 152 | 55 | |
BC012079.1 | - | 1 - 2 | 155 | 55 | |
BQ931455.1 | - | 1 - 2 | 165 | 55 |