Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOCOS
Entrez Gene ID
55034
Gene Name
molybdenum cofactor sulfurase
Gene Aliases
HMCS, MCS, MOS
Chromosome Location
Chr.18: 36187517 - 36268722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017947.2 NP_060417.2 2 - 3 253 75
GenBank mRNA AK000740.1 - 2 - 3 251 75
AK222886.1 - 2 - 3 240 75
BC012079.1 - 2 - 3 243 75
BQ931455.1 - 2 - 3 253 75

View Full Product Details