The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017947.2 | NP_060417.2 | 3 - 4 | 317 | 67 |
GenBank mRNA | AK000740.1 | - | 3 - 4 | 315 | 67 |
AK222886.1 | - | 3 - 4 | 304 | 67 | |
BC012079.1 | - | 3 - 4 | 307 | 67 |