The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017947.2 | NP_060417.2 | 8 - 9 | 1821 | 93 |
GenBank mRNA | AK000740.1 | - | 8 - 9 | 1819 | 93 |
AK222886.1 | - | 8 - 9 | 1808 | 93 | |
AL834481.1 | - | 6 - 7 | 1517 | 93 | |
BC012079.1 | - | 8 - 9 | 1811 | 93 |