Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMURF1
Entrez Gene ID
57154
Gene Name
SMAD specific E3 ubiquitin protein ligase 1
Gene Aliases
-
Chromosome Location
Chr.7: 99027435 - 99144120 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199847.1 NP_001186776.1 11 - 12 1594 114
NM_020429.2 NP_065162.1 12 - 13 1672 114
NM_181349.2 NP_851994.1 11 - 12 1594 114
XM_017012457.1 XP_016867946.1 12 - 13 1670 114
GenBank mRNA AB046845.1 - 12 - 13 1640 114
AF199364.2 - 11 - 12 1227 114
AF464850.1 - 12 - 13 1332 114
BC136804.1 - 11 - 12 1471 114
BC152468.1 - 12 - 13 1640 114

View Full Product Details