The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199847.1 | NP_001186776.1 | 2 - 3 | 432 | 80 |
NM_020429.2 | NP_065162.1 | 2 - 3 | 432 | 80 | |
NM_181349.2 | NP_851994.1 | 2 - 3 | 432 | 80 | |
XM_017012457.1 | XP_016867946.1 | 2 - 3 | 430 | 80 | |
GenBank mRNA | AB046845.1 | - | 2 - 3 | 400 | 80 |
AF199364.2 | - | 2 - 3 | 65 | 80 | |
AF464850.1 | - | 2 - 3 | 92 | 80 | |
BC136804.1 | - | 2 - 3 | 309 | 80 | |
BC144414.1 | - | 2 - 3 | 309 | 80 | |
BC152468.1 | - | 2 - 3 | 400 | 80 | |
DB727777.1 | - | 2 - 3 | 434 | 80 |