Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMURF1
Entrez Gene ID
57154
Gene Name
SMAD specific E3 ubiquitin protein ligase 1
Gene Aliases
-
Chromosome Location
Chr.7: 99027435 - 99144120 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020429.2 NP_065162.1 8 - 9 1148 122
XM_017012457.1 XP_016867946.1 8 - 9 1146 122
GenBank mRNA AB046845.1 - 8 - 9 1116 122
AF464850.1 - 8 - 9 808 122
BC152468.1 - 8 - 9 1116 122

View Full Product Details