Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSTF2
Entrez Gene ID
1478
Gene Name
cleavage stimulation factor subunit 2
Gene Aliases
CstF-64
Chromosome Location
Chr.X: 100820359 - 100841520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306206.1 NP_001293135.1 14 - 15 1842 152
NM_001306209.1 NP_001293138.1 13 - 14 1731 152
NM_001325.2 NP_001316.1 13 - 14 1786 152
GenBank mRNA AK095684.1 - 12 - 13 2119 152
AK300600.1 - 14 - 15 1662 152
AW016125.1 - 1 - 2 152
BC017712.1 - 13 - 14 1755 152
BC033135.1 - 13 - 14 1675 152
EU616679.1 - 14 - 15 1810 152
M85085.1 - 13 - 14 1750 152

View Full Product Details