Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSTF2
Entrez Gene ID
1478
Gene Name
cleavage stimulation factor subunit 2
Gene Aliases
CstF-64
Chromosome Location
Chr.X: 100820359 - 100841520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306206.1 NP_001293135.1 1 - 2 109 94
NM_001306209.1 NP_001293138.1 1 - 2 109 94
NM_001325.2 NP_001316.1 1 - 2 113 94
GenBank mRNA AK095684.1 - 1 - 2 69 94
AK300600.1 - 1 - 2 61 94
BC017712.1 - 1 - 2 82 94
BC033135.1 - 1 - 2 53 94
BT009778.1 - 1 - 2 55 94
BX384743.2 - 1 - 2 109 94
EU616679.1 - 1 - 2 77 94
M85085.1 - 1 - 2 77 94

View Full Product Details