The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253866.1 | NP_001240795.1 | 13 - 14 | 1679 | 125 |
NM_014263.3 | NP_055078.1 | 14 - 15 | 1778 | 125 | |
NM_139312.2 | NP_647473.1 | 15 - 16 | 1949 | 125 | |
XM_011519300.2 | XP_011517602.1 | 14 - 15 | 2232 | 125 | |
GenBank mRNA | AF070656.1 | - | 10 - 11 | 1093 | 125 |
AF151782.1 | - | 15 - 16 | 1793 | 125 | |
AJ132637.1 | - | 14 - 15 | 1751 | 125 | |
AK001259.1 | - | 5 - 6 | 540 | 125 | |
AK291292.1 | - | 14 - 15 | 1707 | 125 | |
AK297973.1 | - | 13 - 14 | 1643 | 125 | |
AK314372.1 | - | 14 - 15 | 1706 | 125 | |
AY358484.1 | - | 14 - 15 | 1704 | 125 | |
BC007795.2 | - | 15 - 16 | 1885 | 125 | |
BC023507.2 | - | 14 - 15 | 1720 | 125 | |
BC024032.2 | - | 14 - 15 | 1725 | 125 |