Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP12
Entrez Gene ID
646486
Gene Name
fatty acid binding protein 12
Gene Aliases
-
Chromosome Location
Chr.8: 81524673 - 81590664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005251285.4 XP_005251342.1 4 - 5 1016 143
XM_006716465.3 XP_006716528.1 4 - 5 999 143
XM_011517577.2 XP_011515879.1 5 - 6 1143 143
GenBank mRNA EU733650.1 - 3 - 4 358 143


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA609025.1 - 1 - 2 Chr. 8 - 81524981 - 81527061 [+] on Build GRCh38 143
GenBankmRNA AI651519.1 - Chr. - - [-] on Build 143
GenBankmRNA BE463892.1 - 1 - 2 Chr. 8 - 81524981 - 81531268 [+] on Build GRCh38 143
GenBankmRNA AI990482.1 - Chr. - - [-] on Build 143
GenBankmRNA AA813151.1 - 1 - 2 Chr. 8 - 81524984 - 81529499 [+] on Build GRCh38 143
GenBankmRNA AX785174.1 - 1 - 2 Chr. 8 - 81524981 - 81531268 [+] on Build GRCh38 143
GenBankmRNA AA812713.1 - 1 - 2 Chr. 8 - 81524987 - 81529516 [+] on Build GRCh38 143


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream fatty acid binding protein 4 FABP4 A-FABP, AFABP, ALBP, HEL-S-104, aP2 -41.7kb Anti-sense Chr. - 81478497 - 81483238 on Build GRCh38
Downstream inositol monophosphatase 1 IMPA1 IMP, IMPA 129.8kb Anti-sense Chr. - 81656916 - 81686354 on Build GRCh38

View Full Product Details