The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031748.3 | NP_001026918.2 | 9 - 10 | 1454 | 85 |
NM_001319247.1 | NP_001306176.1 | 9 - 10 | 1454 | 85 | |
XM_005268806.3 | XP_005268863.1 | 8 - 9 | 1264 | 85 | |
XM_011538231.2 | XP_011536533.1 | 8 - 9 | 1491 | 85 | |
GenBank mRNA | BC038754.1 | - | 9 - 10 | 1451 | 85 |
BC048120.1 | - | 9 - 10 | 1438 | 85 |