The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014248.3 | NP_055063.1 | 4 - 5 | 369 | 108 |
GenBank mRNA | AF085906.1 | - | 4 - 5 | 298 | 108 |
AK090764.1 | - | 1 - 2 | 1687 | 108 | |
BC001466.2 | - | 4 - 5 | 322 | 108 | |
BC017370.1 | - | 4 - 5 | 339 | 108 | |
CR456560.1 | - | 4 - 5 | 335 | 108 | |
DN602016.1 | - | 1 - 2 | 47 | 108 |