The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288969.1 | NP_001275898.1 | 1 - 2 | 231 | 113 |
NM_001288970.1 | NP_001275899.1 | 1 - 2 | 231 | 113 | |
NM_001288972.1 | NP_001275901.1 | 1 - 2 | 231 | 113 | |
NM_025149.5 | NP_079425.3 | 1 - 2 | 231 | 113 | |
XM_006722110.3 | XP_006722173.1 | 1 - 2 | 171 | 113 | |
XM_011525294.1 | XP_011523596.1 | 1 - 2 | 171 | 113 | |
GenBank mRNA | AK024573.1 | - | 1 - 2 | 138 | 113 |
AK225044.1 | - | 1 - 2 | 138 | 113 | |
AK225561.1 | - | 1 - 2 | 137 | 113 | |
AK295258.1 | - | 1 - 2 | 139 | 113 | |
AK300145.1 | - | 1 - 2 | 139 | 113 | |
AY358660.1 | - | 1 - 2 | 167 | 113 | |
BC012053.1 | - | 1 - 2 | 144 | 113 | |
BC014123.2 | - | 1 - 2 | 151 | 113 | |
DC296100.1 | - | 1 - 2 | 171 | 113 |