The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001618.3 | NP_001609.2 | 12 - 13 | 1911 | 92 |
GenBank mRNA | AK225654.1 | - | 12 - 13 | 1876 | 92 |
AK303340.1 | - | 12 - 13 | 1838 | 92 | |
AK312339.1 | - | 12 - 13 | 1827 | 92 | |
BC037545.1 | - | 12 - 13 | 1884 | 92 | |
J03473.1 | - | 12 - 13 | 1835 | 92 | |
M17081.1 | - | 3 - 4 | 420 | 92 | |
M18112.1 | - | 12 - 13 | 1879 | 92 | |
M32721.1 | - | 12 - 13 | 1899 | 92 |