The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318066.1 | NP_001304995.1 | 3 - 4 | 547 | 86 |
NM_014822.3 | NP_055637.2 | 3 - 4 | 547 | 86 | |
XM_005263379.2 | XP_005263436.1 | 3 - 4 | 421 | 86 | |
GenBank mRNA | AB018298.1 | - | 3 - 4 | 450 | 86 |
AF130464.2 | - | 2 - 3 | 250 | 86 | |
AK291765.1 | - | 3 - 4 | 447 | 86 | |
BC035761.1 | - | 3 - 4 | 469 | 86 | |
BC037736.1 | - | 4 - 5 | 591 | 86 |