The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002253.2 | NP_002244.1 | 11 - 12 | 1839 | 110 |
GenBank mRNA | AB209901.1 | - | 11 - 12 | 1824 | 110 |
AF035121.1 | - | 11 - 12 | 1840 | 110 | |
AF063658.1 | - | 11 - 12 | 1537 | 110 | |
BC131822.1 | - | 11 - 12 | 1599 | 110 | |
EU826563.1 | - | 11 - 12 | 1537 | 110 | |
FJ899739.1 | - | 11 - 12 | 1537 | 110 | |
L04947.1 | - | 11 - 12 | 1531 | 110 | |
X61656.1 | - | 11 - 12 | 1531 | 110 |