The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002253.2 | NP_002244.1 | 25 - 26 | 3707 | 90 |
GenBank mRNA | AB209901.1 | - | 25 - 26 | 3692 | 90 |
AF035121.1 | - | 25 - 26 | 3708 | 90 | |
AF063658.1 | - | 25 - 26 | 3405 | 90 | |
BC131822.1 | - | 25 - 26 | 3467 | 90 | |
L04947.1 | - | 25 - 26 | 3399 | 90 | |
X61656.1 | - | 25 - 26 | 3399 | 90 |