The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018725.3 | NP_061195.2 | 3 - 4 | 269 | 120 |
XM_005265310.4 | XP_005265367.1 | 3 - 4 | 269 | 120 | |
XM_005265311.4 | XP_005265368.1 | 3 - 4 | 294 | 120 | |
XM_005265312.4 | XP_005265369.1 | 3 - 4 | 325 | 120 | |
XM_011533941.2 | XP_011532243.1 | 3 - 4 | 269 | 120 | |
XM_017006807.1 | XP_016862296.1 | 3 - 4 | 269 | 120 | |
GenBank mRNA | AF208110.1 | - | 3 - 4 | 235 | 120 |
AF208111.1 | - | 3 - 4 | 235 | 120 | |
AF212365.1 | - | 3 - 4 | 239 | 120 | |
AF250309.1 | - | 3 - 4 | 230 | 120 | |
AK307749.1 | - | 3 - 4 | 251 | 120 | |
AY359122.1 | - | 3 - 4 | 236 | 120 | |
BC000980.1 | - | 3 - 4 | 239 | 120 | |
DA093617.1 | - | 3 - 4 | 269 | 120 |