The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024658.3 | NP_078934.3 | 16 - 17 | 1764 | 83 |
NR_051979.1 | 16 - 17 | 1764 | 83 | ||
GenBank mRNA | AB208897.1 | - | 16 - 17 | 1653 | 83 |
AF411122.1 | - | 16 - 17 | 1635 | 83 | |
AK026991.1 | - | 9 - 10 | 939 | 83 | |
AK056527.1 | - | 16 - 17 | 1685 | 83 | |
AK056702.1 | - | 16 - 17 | 1611 | 83 | |
AK057044.1 | - | 15 - 16 | 1745 | 83 | |
AK057277.1 | - | 16 - 17 | 1668 | 83 | |
AK094107.1 | - | 16 - 17 | 1612 | 83 | |
AK094897.1 | - | 15 - 16 | 1771 | 83 | |
AK095306.1 | - | 16 - 17 | 1612 | 83 | |
AK302097.1 | - | 10 - 11 | 804 | 83 | |
AK314305.1 | - | 15 - 16 | 1529 | 83 | |
BC003690.1 | - | 7 - 8 | 751 | 83 | |
BC110804.1 | - | 16 - 17 | 1666 | 83 | |
BC136759.1 | - | 16 - 17 | 1715 | 83 | |
BC144399.1 | - | 16 - 17 | 1715 | 83 | |
BX248267.1 | - | 16 - 17 | 1646 | 83 | |
CR457367.1 | - | 7 - 8 | 661 | 83 |