The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024658.3 | NP_078934.3 | 17 - 18 | 1891 | 73 |
NR_051979.1 | 17 - 18 | 1891 | 73 | ||
GenBank mRNA | AB208897.1 | - | 17 - 18 | 1780 | 73 |
AF411122.1 | - | 17 - 18 | 1762 | 73 | |
AK026991.1 | - | 10 - 11 | 1066 | 73 | |
AK056527.1 | - | 17 - 18 | 1812 | 73 | |
AK056702.1 | - | 17 - 18 | 1738 | 73 | |
AK057044.1 | - | 16 - 17 | 1872 | 73 | |
AK057277.1 | - | 17 - 18 | 1795 | 73 | |
AK074743.1 | - | 2 - 3 | 150 | 73 | |
AK094107.1 | - | 17 - 18 | 1739 | 73 | |
AK094897.1 | - | 16 - 17 | 1898 | 73 | |
AK095306.1 | - | 17 - 18 | 1739 | 73 | |
AK302097.1 | - | 11 - 12 | 931 | 73 | |
AK314305.1 | - | 16 - 17 | 1656 | 73 | |
BC003690.1 | - | 8 - 9 | 878 | 73 | |
BC110804.1 | - | 17 - 18 | 1793 | 73 | |
BC136759.1 | - | 17 - 18 | 1842 | 73 | |
BC144399.1 | - | 17 - 18 | 1842 | 73 | |
BX248267.1 | - | 17 - 18 | 1773 | 73 | |
CR457367.1 | - | 8 - 9 | 788 | 73 |