Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IPO4
Entrez Gene ID
79711
Gene Name
importin 4
Gene Aliases
Imp4
Chromosome Location
Chr.14: 24180219 - 24188918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024658.3 NP_078934.3 29 - 30 3243 64
NR_051979.1 29 - 30 3263 64
GenBank mRNA AB208897.1 - 29 - 30 3132 64
AF411122.1 - 29 - 30 3114 64
AK026991.1 - 22 - 23 2418 64
AK027871.1 - 11 - 12 1211 64
AK056527.1 - 29 - 30 3184 64
AK056702.1 - 29 - 30 3090 64
AK057044.1 - 28 - 29 3224 64
AK057277.1 - 29 - 30 3147 64
AK094107.1 - 29 - 30 3091 64
AK094897.1 - 28 - 29 3250 64
AK095306.1 - 29 - 30 3091 64
AK302097.1 - 23 - 24 2301 64
AK314305.1 - 27 - 28 3115 64
BC003690.1 - 20 - 21 2230 64
BC110804.1 - 29 - 30 3145 64
BC136759.1 - 29 - 30 3194 64
BC144399.1 - 29 - 30 3214 64
BX248267.1 - 29 - 30 3125 64
CR457367.1 - 20 - 21 2140 64

View Full Product Details