The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024658.3 | NP_078934.3 | 8 - 9 | 883 | 60 |
NR_051979.1 | 8 - 9 | 883 | 60 | ||
GenBank mRNA | AF411122.1 | - | 8 - 9 | 754 | 60 |
AK026991.1 | - | 1 - 2 | 58 | 60 | |
AK056527.1 | - | 8 - 9 | 804 | 60 | |
AK056702.1 | - | 8 - 9 | 730 | 60 | |
AK057044.1 | - | 7 - 8 | 841 | 60 | |
AK057277.1 | - | 8 - 9 | 787 | 60 | |
AK094107.1 | - | 8 - 9 | 732 | 60 | |
AK094897.1 | - | 7 - 8 | 890 | 60 | |
AK095306.1 | - | 8 - 9 | 731 | 60 | |
AK314305.1 | - | 8 - 9 | 762 | 60 | |
BC110804.1 | - | 8 - 9 | 785 | 60 | |
BC136759.1 | - | 8 - 9 | 834 | 60 | |
BC144399.1 | - | 8 - 9 | 834 | 60 | |
BX248267.1 | - | 8 - 9 | 765 | 60 |