The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031847.2 | NP_001027017.1 | 10 - 11 | 1330 | 130 |
NM_001876.3 | NP_001867.2 | 10 - 11 | 1330 | 130 | |
XM_005273762.2 | XP_005273819.1 | 10 - 11 | 1306 | 130 | |
XM_005273763.1 | XP_005273820.1 | 10 - 11 | 1306 | 130 | |
XM_017017220.1 | XP_016872709.1 | 10 - 11 | 1439 | 130 | |
GenBank mRNA | AK025307.1 | - | 5 - 6 | 616 | 130 |
AK172798.1 | - | 10 - 11 | 1316 | 130 | |
AK309464.1 | - | 10 - 11 | 1306 | 130 | |
AK314301.1 | - | 10 - 11 | 1269 | 130 | |
BC000185.1 | - | 10 - 11 | 1205 | 130 | |
BT009791.1 | - | 9 - 10 | 1160 | 130 | |
L39211.1 | - | 10 - 11 | 1236 | 130 |