Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPT1A
Entrez Gene ID
1374
Gene Name
carnitine palmitoyltransferase 1A
Gene Aliases
CPT1, CPT1-L, L-CPT1
Chromosome Location
Chr.11: 68754620 - 68844410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031847.2 NP_001027017.1 7 - 8 943 67
NM_001876.3 NP_001867.2 7 - 8 943 67
XM_005273762.2 XP_005273819.1 7 - 8 919 67
XM_005273763.1 XP_005273820.1 7 - 8 919 67
XM_017017220.1 XP_016872709.1 7 - 8 1052 67
GenBank mRNA AK025307.1 - 2 - 3 229 67
AK172798.1 - 7 - 8 929 67
AK309464.1 - 7 - 8 919 67
AK314301.1 - 7 - 8 882 67
BC000185.1 - 7 - 8 818 67
BT009791.1 - 6 - 7 773 67
L39211.1 - 7 - 8 849 67

View Full Product Details