The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031847.2 | NP_001027017.1 | 8 - 9 | 1046 | 76 |
NM_001876.3 | NP_001867.2 | 8 - 9 | 1046 | 76 | |
XM_005273762.2 | XP_005273819.1 | 8 - 9 | 1022 | 76 | |
XM_005273763.1 | XP_005273820.1 | 8 - 9 | 1022 | 76 | |
XM_017017220.1 | XP_016872709.1 | 8 - 9 | 1155 | 76 | |
GenBank mRNA | AK025307.1 | - | 3 - 4 | 332 | 76 |
AK172798.1 | - | 8 - 9 | 1032 | 76 | |
AK309464.1 | - | 8 - 9 | 1022 | 76 | |
AK314301.1 | - | 8 - 9 | 985 | 76 | |
BC000185.1 | - | 8 - 9 | 921 | 76 | |
BT009791.1 | - | 7 - 8 | 876 | 76 | |
L39211.1 | - | 8 - 9 | 952 | 76 |