The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139514.1 | NP_001132986.1 | 4 - 5 | 1063 | 93 |
NM_001139515.1 | NP_001132987.1 | 5 - 6 | 488 | 93 | |
NM_053281.3 | NP_444511.1 | 5 - 6 | 1102 | 93 | |
XM_011530846.2 | XP_011529148.1 | 5 - 6 | 1101 | 93 | |
XM_011530847.2 | XP_011529149.1 | 5 - 6 | 1101 | 93 | |
XM_011530848.2 | XP_011529150.1 | 5 - 6 | 1101 | 93 | |
XM_017029254.1 | XP_016884743.1 | 4 - 5 | 1062 | 93 | |
XM_017029255.1 | XP_016884744.1 | 4 - 5 | 1062 | 93 | |
XM_017029256.1 | XP_016884745.1 | 4 - 5 | 1062 | 93 | |
GenBank mRNA | AF428101.1 | - | 5 - 6 | 932 | 93 |
AK055953.1 | - | 4 - 5 | 449 | 93 | |
AK091842.1 | - | 4 - 5 | 1056 | 93 | |
AK290596.1 | - | 5 - 6 | 1102 | 93 | |
AK298787.1 | - | 5 - 6 | 488 | 93 | |
BC114950.1 | - | 5 - 6 | 1011 | 93 |