Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DACH2
Entrez Gene ID
117154
Gene Name
dachshund family transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.X: 86148451 - 86832602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139514.1 NP_001132986.1 6 - 7 1370 90
NM_001139515.1 NP_001132987.1 7 - 8 795 90
NM_053281.3 NP_444511.1 7 - 8 1409 90
XM_011530846.2 XP_011529148.1 7 - 8 1408 90
XM_011530847.2 XP_011529149.1 7 - 8 1408 90
XM_011530848.2 XP_011529150.1 7 - 8 1408 90
XM_017029254.1 XP_016884743.1 6 - 7 1369 90
XM_017029255.1 XP_016884744.1 6 - 7 1369 90
XM_017029256.1 XP_016884745.1 6 - 7 1369 90
GenBank mRNA AF428101.1 - 7 - 8 1239 90
AK055953.1 - 6 - 7 756 90
AK091842.1 - 6 - 7 1363 90
AK290596.1 - 7 - 8 1409 90
AK294498.1 - 2 - 3 204 90
AK295648.1 - 2 - 3 205 90
AK298787.1 - 7 - 8 795 90
BC114950.1 - 7 - 8 1318 90

View Full Product Details