The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005865.3 | NP_005856.1 | 2 - 3 | 250 | 70 |
XM_017010161.1 | XP_016865650.1 | 2 - 3 | 250 | 70 | |
XM_017010162.1 | XP_016865651.1 | 2 - 3 | 250 | 70 | |
XM_017010163.1 | XP_016865652.1 | 2 - 3 | 250 | 70 | |
XM_017010164.1 | XP_016865653.1 | 2 - 3 | 250 | 70 | |
GenBank mRNA | AF052514.1 | - | 2 - 3 | 247 | 70 |
AK303106.1 | - | 2 - 3 | 242 | 70 | |
AK314037.1 | - | 2 - 3 | 250 | 70 |