Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WAPL
Entrez Gene ID
23063
Gene Name
WAPL cohesin release factor
Gene Aliases
FOE, KIAA0261, WAPAL
Chromosome Location
Chr.10: 86435256 - 86521816 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318328.1 NP_001305257.1 5 - 6 2163 98
NM_015045.3 NP_055860.1 5 - 6 2181 98
XM_011539548.1 XP_011537850.1 5 - 6 2231 98
XM_017015981.1 XP_016871470.1 2 - 3 1768 98
GenBank mRNA AB065003.1 - 5 - 6 2184 98
AF479418.1 - 6 - 7 1859 98
AK290138.1 - 5 - 6 2142 98
BC140858.1 - 5 - 6 1887 98
BC150269.1 - 6 - 7 2032 98
D87450.1 - 6 - 7 2032 98

View Full Product Details