The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318328.1 | NP_001305257.1 | 8 - 9 | 2566 | 84 |
NM_015045.3 | NP_055860.1 | 8 - 9 | 2584 | 84 | |
XM_011539548.1 | XP_011537850.1 | 8 - 9 | 2634 | 84 | |
XM_017015979.1 | XP_016871468.1 | 7 - 8 | 2481 | 84 | |
XM_017015980.1 | XP_016871469.1 | 7 - 8 | 2424 | 84 | |
XM_017015981.1 | XP_016871470.1 | 5 - 6 | 2171 | 84 | |
GenBank mRNA | AB065003.1 | - | 8 - 9 | 2587 | 84 |
AF479418.1 | - | 9 - 10 | 2262 | 84 | |
AK027005.1 | - | 1 - 2 | 307 | 84 | |
AK290138.1 | - | 8 - 9 | 2545 | 84 | |
BC140858.1 | - | 8 - 9 | 2290 | 84 | |
BC150269.1 | - | 9 - 10 | 2435 | 84 | |
D87450.1 | - | 9 - 10 | 2435 | 84 |