Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTGFR
Entrez Gene ID
5737
Gene Name
prostaglandin F receptor
Gene Aliases
FP
Chromosome Location
Chr.1: 78490974 - 78540701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000959.3 NP_000950.1 1 - 2 192 89
NM_001039585.1 NP_001034674.1 1 - 2 192 89
XM_017001873.1 XP_016857362.1 1 - 2 261 89
GenBank mRNA AF004021.1 - 1 - 2 77 89
AK292845.1 - 1 - 2 176 89
AK312816.1 - 1 - 2 176 89
AY485530.1 - 1 - 2 163 89
BC112965.2 - 1 - 2 103 89
CD013876.1 - 1 - 2 196 89
L24470.1 - 1 - 2 163 89

View Full Product Details