Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP1
Entrez Gene ID
65055
Gene Name
receptor accessory protein 1
Gene Aliases
C2orf23, HMN5B, SPG31, Yip2a
Chromosome Location
Chr.2: 86213993 - 86338083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164730.1 NP_001158202.1 3 - 4 205 84
NM_001164731.1 NP_001158203.1 2 - 3 248 84
NM_022912.2 NP_075063.1 3 - 4 321 84
XM_005264502.1 XP_005264559.1 3 - 4 321 84
XM_005264504.1 XP_005264561.1 3 - 4 239 84
XM_011533044.1 XP_011531346.1 3 - 4 183 84
XM_011533045.2 XP_011531347.1 3 - 4 168 84
XM_017004725.1 XP_016860214.1 3 - 4 489 84
XM_017004726.1 XP_016860215.1 3 - 4 489 84
XM_017004727.1 XP_016860216.1 3 - 4 489 84
GenBank mRNA AK023172.1 - 3 - 4 321 84
AK294300.1 - 3 - 4 302 84
AK297287.1 - 3 - 4 205 84
AK297359.1 - 3 - 4 293 84
AK297373.1 - 4 - 5 433 84
AK299334.1 - 2 - 3 220 84
AY562239.1 - 3 - 4 177 84
BC064846.1 - 3 - 4 283 84
CR457301.1 - 3 - 4 177 84
DA495719.1 - 3 - 4 205 84

View Full Product Details