Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DOPEY1
- Entrez Gene ID
-
23033
- Gene Name
- dopey family member 1
- Gene Aliases
-
DOP1, KIAA1117, dJ202D23.2
- Chromosome Location
-
Chr.6: 83067650 - 83171351 on Build GRCh38
View Full Product Details