The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031701.2 | NP_001026871.1 | 12 - 13 | 1367 | 68 |
XM_011538476.2 | XP_011536778.1 | 9 - 10 | 1124 | 68 | |
GenBank mRNA | AB032773.1 | - | 12 - 13 | 1177 | 68 |
AK002128.1 | - | 1 - 2 | 48 | 68 | |
BC052293.1 | - | 12 - 13 | 1367 | 68 |