The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016262.4 | NP_057346.1 | 2 - 3 | 217 | 75 |
XM_011535875.2 | XP_011534177.1 | 2 - 3 | 198 | 75 | |
XM_017010918.1 | XP_016866407.1 | 2 - 3 | 162 | 75 | |
GenBank mRNA | AF201334.1 | - | 2 - 3 | 103 | 75 |
AK026242.1 | - | 2 - 3 | 182 | 75 | |
BC025405.2 | - | 2 - 3 | 122 | 75 | |
BC031101.1 | - | 2 - 3 | 170 | 75 | |
DA160210.1 | - | 2 - 3 | 217 | 75 |