The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142353.2 | NP_001135825.1 | 5 - 6 | 811 | 114 |
NM_001142354.2 | NP_001135826.1 | 5 - 6 | 811 | 114 | |
NM_001289968.1 | NP_001276897.1 | 6 - 7 | 865 | 114 | |
NM_001289969.1 | NP_001276898.1 | 6 - 7 | 930 | 114 | |
NM_021132.3 | NP_066955.1 | 5 - 6 | 811 | 114 | |
XM_005269944.1 | XP_005270001.1 | 5 - 6 | 811 | 114 | |
XM_017016379.1 | XP_016871868.1 | 5 - 6 | 811 | 114 | |
GenBank mRNA | AJ488506.1 | - | 5 - 6 | 748 | 114 |
AK301582.1 | - | 6 - 7 | 837 | 114 | |
BC028049.1 | - | 5 - 6 | 799 | 114 | |
BC035464.1 | - | 5 - 6 | 769 | 114 | |
M29550.1 | - | 6 - 7 | 837 | 114 | |
M29551.1 | - | 5 - 6 | 792 | 114 |