Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA12
Entrez Gene ID
26154
Gene Name
ATP binding cassette subfamily A member 12
Gene Aliases
ARCI4A, ARCI4B, ICR2B, LI2
Chromosome Location
Chr.2: 214931542 - 215138591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015657.3 NP_056472.2 34 - 35 5445 67
NM_173076.2 NP_775099.2 42 - 43 6460 67
NR_103740.1 44 - 45 6540 67
XM_011510951.2 XP_011509253.1 42 - 43 6632 67
GenBank mRNA AF418105.1 - 37 - 38 5582 67
AK123578.1 - 9 - 10 1000 67
AY033486.1 - 34 - 35 67

View Full Product Details